Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C17orf74 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569937
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569937 Supplier Invitrogen™ Supplier No. PA569937
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Horse: 77%; Human: 100%.

C17orf74 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C17orf74
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SPEM2
Gene Accession No. Q0P670
Gene Alias C17orf74; chromosome 17 open reading frame 74; SPEM2; uncharacterized protein C17orf74; Uncharacterized protein SPEM2
Gene Symbols SPEM2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human C17orf74.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201243
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.