Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C17orf74 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569938
Description
This target displays homology in the following species: Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
C17orf74 is a protein coding gene.
Specifications
C17orf74 | |
Polyclonal | |
Unconjugated | |
SPEM2 | |
C17orf74; chromosome 17 open reading frame 74; SPEM2; uncharacterized protein C17orf74; Uncharacterized protein SPEM2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
201243 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q0P670 | |
SPEM2 | |
synthetic peptide directed towards the C-terminal region of human C17orf74. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction