Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C17orf75 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS9646R

Description
The function of this protein remains unknown.Specifications
C17orf75 | |
Polyclonal | |
Unconjugated | |
C17orf75 | |
5730455P16Rik; C17orf75; chromosome 17 open reading frame 75; NJMU-R1; Protein Njmu-R1; RGD1310429; RIKEN cDNA 5730455P16 gene; sensitization to ricin complex subunit 2; similar to Protein Njmu-R1; spermatogenesis-related protein; SRI2 | |
Rabbit | |
Protein A | |
RUO | |
64149 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9HAS0 | |
C17orf75 | |
KLH conjugated synthetic peptide derived from human C17orf75. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction