Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C17orf80 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145055
Description
Immunogen sequence: KLVVDKPEQT VKTFPLPAVG LERAATTKAD KDIKNPIQPS FKMLKNTKPM TTFQEETKAQ FYASEKTSPK RELAKDLPKS GESRCNP Antigen sequence identity: Mouse 37%; Rat 33%.

Specifications
C17orf80 | |
Polyclonal | |
Unconjugated | |
C17orf80 | |
C17orf80; Cell migration-inducing gene 3 protein; chromosome 17 open reading frame 80; HLC8; HLC-8; Human lung cancer oncogene 8 protein; lung cancer-related protein 8; MIG3; SPEP1; sperm-expressed protein 1; uncharacterized protein C17orf80 | |
Rabbit | |
Affinity chromatography | |
RUO | |
55028 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Western Blot, Immunocytochemistry | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BSJ5 | |
C17orf80 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 80-166. Recombinant protein control fragment (Product #RP-110138). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction