Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C17orf81 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544141
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544141 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544141 Supplier Invitrogen™ Supplier No. PA544141
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FSILPDFSLD LQEGPSVESQ PYSDPHIPPV SKNAKARTRK CSLVSGHGRE Sequence homology: Human: 100%.

C17orf81 belongs to the ELP5 family. C17orf81 may be involved in TP53-mediated transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C17orf81
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ELP5
Gene Accession No. Q8TE02
Gene Alias C17orf81; dermal papilla derived protein 6; Dermal papilla-derived protein 6; Dermal papilla-derived protein 6 homolog; Derp6; elongator acetyltransferase complex subunit 5; elongator complex protein 5; Elp5; HSPC002; MST071; MSTP071; Rai12; retinoic acid induced 12; retinoic acid-induced protein 12; S-phase 2 protein
Gene Symbols ELP5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human C17orf81.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23587
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.