Learn More
Invitrogen™ C17orf81 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120311
Description
Positive test controls include: U-251MG, LO2, A375, Mouse brain, Mouse skeletal muscle, Mouse testis. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: MLDSLLALGG LVLLRDSVEW EGRSLLKALV KKSALCGEQV HILGCEVSEE EFREGFDSDI NNRLVYHDFF RDPLNWSKTE EAFPGGPLGA LRAMCKRTDP VPVTIALDSL SWLLLRLPCT TLCQVLHAVS HQDSCPGDSS SVGKVSVLGL LHEELHGPGP VGALSSLAQT EVTLGGTMGQ ASAHILCRRP RQRPTDQTQW FSILPDFSLD LQEGPSVESQ PYSDPHIPPV DPTTHLTFNL HLSKKEREAR DSLILPFQFS SEKQQALLRP RPGQATSHIF YEPDAYDDLD QEDPDDDLDI.
C17orf81 belongs to the ELP5 family. C17orf81 may be involved in TP53-mediated transcriptional regulation.
Specifications
C17orf81 | |
Polyclonal | |
Unconjugated | |
ELP5 | |
C17orf81; dermal papilla derived protein 6; Dermal papilla-derived protein 6; Dermal papilla-derived protein 6 homolog; Derp6; elongator acetyltransferase complex subunit 5; elongator complex protein 5; Elp5; HSPC002; MST071; MSTP071; Rai12; retinoic acid induced 12; retinoic acid-induced protein 12; S-phase 2 protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
23587, 54351 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.63 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8TE02, Q99L85 | |
ELP5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 17-316 of human ELP5 (NP_056177.3). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.