Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C17orf81 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 101621AP150UL

Description
C17orf81 belongs to the ELP5 family. C17orf81 may be involved in TP53-mediated transcriptional regulation.Specifications
C17orf81 | |
Polyclonal | |
Unconjugated | |
ELP5 | |
C17orf81, DERP6, ELP5, HSPC002, MST071, MSTP071, S phase 2 protein, S-phase 2 protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23587 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TE02 | |
ELP5 | |
C17orf81 Fusion Protein Ag0194 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction