Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C17orf82 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989229
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9229 Supplier Bioss Supplier No. BS9649R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The specific function of this protein remains unknown.

Specifications

Antigen C17orf82
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C17orf82
Gene Accession No. Q86X59
Gene Alias C17orf82; chromosome 17 open reading frame 82; putative uncharacterized protein C17orf82
Gene Symbols C17orf82
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C17orf82.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388407
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.