Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C17orf89 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571013 Supplier Thermo Scientific Supplier No. PA571013
Add to Cart
Add to Cart

C17orf89 Polyclonal Antibody

C17orf89 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen C17orf89
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NDUFAF8
Gene Accession No. A1L188
Gene Alias 1810043H04Rik; C17orf89; C9H17orf89; chromosome 17 open reading frame 89; hypothetical protein LOC690871; LOC690871; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 8; NADH:ubiquinone oxidoreductase complex assembly factor 8; Ndufaf8; RIKEN cDNA 1810043H04 gene; uncharacterized protein C17orf89; uncharacterized protein C17orf89 homolog
Gene Symbols NDUFAF8
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human C17orf89.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284184
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.