Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C17orf96 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565930
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565930 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565930 Supplier Invitrogen™ Supplier No. PA565930
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ETLCPAPRLAV PASPRGSPCS PTPRKPCRGT QEFSPL Highest antigen sequence identity to the following orthologs - mouse 89%, rat 92%.

Elongin BC and Polycomb repressive complex 2-associated protein (EPOP) also know as C17ORF96 is a scaffold protein that serves as a bridging partner between the PRC2/EED-EZH2 complex and the elongin BC complex. It is required to fine tune the transcriptional status of polycomb group (PcG) target genes in embryonic stem cells (ESCs). C17ORF96 also plays a key role in genomic regions that display both active and repressive chromatin properties in pluripotent stem cells by sustaining low level expression of PcG target genes. It acts by recruiting the elongin BC complex, thereby restricting excessive activity of the PRC2/EED-EZH2 complex. C17ORF96 also has interacts with USP7 and promotes deubiquitination of H2B at promoter sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C17orf96
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EPOP
Gene Accession No. A6NHQ4
Gene Alias AA409164; AI413509; AW539173; C17orf96; C9H17orf96; chromosome 17 open reading frame 96; CUNH17orf96; E13; E130012A19Rik; elongin BC and polycomb repressive complex 2 associated protein; elongin BC and Polycomb repressive complex 2-associated protein; Embryonic stem cell-specific PRC2 subunit p48; EPOP; ES cell-specific PRC2 subunit p48; esPRC2p48; hypothetical protein LOC691153; LOC691153; proline rich 28; Proline-rich protein 28; PRR28; RIKEN cDNA E130012A19 gene; uncharacterized protein C17orf96; uncharacterized protein C17orf96 homolog; Uncharacterized protein ENSP00000317905
Gene Symbols EPOP
Host Species Rabbit
Immunogen Recombinant Human C17orf96. Recombinant protein control fragment (Product #RP-106610).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100170841
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.