Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C17orf97 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570016
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 86%; Human: 100%; Mouse: 100%; Pig: 86%; Rabbit: 93%; Rat: 100%.
C17orf97 gene ontology annotations related to this gene include protein arginylation.
Specifications
C17orf97 | |
Polyclonal | |
Unconjugated | |
C17orf97 | |
C17orf97; chromosome 17 open reading frame 97; CK20; LIAT1; Ligand of ATE1 protein; protein LIAT1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
400566 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6ZQX7 | |
C17orf97 | |
synthetic peptide directed towards the N terminal region of human C17orf97. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction