Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C17orf99 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556224
Description
Immunogen sequence: FSFLPSQTSD WFWCQAANNA NVQHSALTVV PPGGDQKMED WQGPLESPIL ALPLYRSTRR LSEEEFGGFR IGNGEVRGRK AAAM Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 23%.
Probable B cell-associated cytokine that plays a role in the regulation of humoral immune responses. Involved in lymphocyte B cell development and immunoglobulin/IgA production.
Specifications
C17orf99 | |
Polyclonal | |
Unconjugated | |
C17orf99 | |
C17orf99; chromosome 17 open reading frame 99; GLPG464; IL40; IL-40; Interleukin-40; Protein IL-40; uncharacterized protein C17orf99; UNQ464; UNQ464/PRO809 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100141515 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UX52 | |
C17orf99 | |
Recombinant protein corresponding to Human C17orf99. Recombinant protein control fragment (Product #RP-93530). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction