Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C18orf23 Monoclonal Antibody (3A1)
GREENER_CHOICE

Catalog No. PIMA523079
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523079 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523079 Supplier Invitrogen™ Supplier No. MA523079
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: VSHPRRSQER VSVHPHRLHP SFDFGQLQTP QPRYLAEGTD WDLSVDAGLS PAQFQVRPIP QHYQHYLATP RMHHFPRNSS STQMVVHEIR NYPYPQLHF.

C18orf23 (NP_689683, 122 a.a. to approximately 220 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C18orf23
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3A1
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Host Species Mouse
Immunogen C18orf23 (NP_689683, 122 a.a. ~ 220 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.