Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C18orf25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113951
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113951 Supplier Invitrogen™ Supplier No. PA5113951
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STSSSDDDEE VSGSSKTITA EIPGHLDPGF LASDKTSAGN APLNEEINIA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

The exact function of C18orf25 remains unknown. There are two named isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C18orf25
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C18orf25
Gene Accession No. Q96B23
Gene Alias 8030462N17Rik; Akd2; Ark2N; arkadia 2; ARKadia-like 1; ARKadia-like protein 1; ARKL1; C18orf25; chromosome 18 open reading frame 25; cr025; RIKEN cDNA 8030462N17 gene; RNF111L1; Uncharacterized protein C18orf25; uncharacterized protein C18orf25 homolog
Gene Symbols C18orf25
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C18orf25.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147339
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.