Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C18orf25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562379
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562379 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562379 Supplier Invitrogen™ Supplier No. PA562379
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESQLASTESD KPTTGRVYES DSSNHCMLSP SSSGHLADSD TLSSAEENEP SQAETAVEGD PSGVSGATVG RKSRRSRSES ETSTMA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 95%.

The exact function of C18orf25 remains unknown. There are two named isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C18orf25
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C18orf25
Gene Accession No. Q96B23
Gene Alias 8030462N17Rik; Akd2; Ark2N; arkadia 2; ARKadia-like 1; ARKadia-like protein 1; ARKL1; C18orf25; chromosome 18 open reading frame 25; cr025; RIKEN cDNA 8030462N17 gene; RNF111L1; Uncharacterized protein C18orf25; uncharacterized protein C18orf25 homolog
Gene Symbols C18orf25
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C18orf25. Recombinant protein control fragment (Product #RP-101356).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147339
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.