Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C18orf42 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572891
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572891 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572891 Supplier Thermo Scientific Supplier No. PA572891
Add to Cart
Add to Cart

C18orf42 Polyclonal Antibody

C18orf42 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen C18orf42
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene AKAIN1
Gene Accession No. P0CW23
Gene Alias A kinase (PRKA) anchor inhibitor 1; AKAIN1; A-kinase anchor protein C18orf42; A-kinase anchor protein inhibitor 1; C18orf42
Gene Symbols AKAIN1
Host Species Rabbit
Immunogen C18orf42 antibody for a 16 amino acid peptide from near the center of human C18orf42. The immunogen is within the last 50 amino acids of C18orf42.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 642597
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.