Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C18orf42 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561944 Supplier Invitrogen™ Supplier No. PA561944
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FWLGEKPGNE PEEVKLQNAS KQIVQNAILQ AVQQVSQESQ RREERISDNR DHIQLGVGEL TKKHEKK Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 75%.

Protein kinase A (PKA)-binding protein. Binds to type II regulatory subunits of protein kinase A (PKA) and may block the A-kinase anchoring protein (AKAP)-mediated subcellular localization of PKA (PubMed:25653177).
TRUSTED_SUSTAINABILITY

Specifications

Antigen C18orf42
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AKAIN1
Gene Accession No. P0CW23
Gene Alias A kinase (PRKA) anchor inhibitor 1; AKAIN1; A-kinase anchor protein C18orf42; A-kinase anchor protein inhibitor 1; C18orf42
Gene Symbols AKAIN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C18orf42. Recombinant protein control fragment (Product #RP-100684).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 642597
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.