Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C18orf54 Monoclonal Antibody (OTI2B2), TrueMAB™, OriGene
SDP

Catalog No. 501682052
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2052 Supplier OriGene Supplier No. TA808319
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Might play a role in cell proliferation.

Specifications

Antigen C18orf54
Applications Western Blot
Classification Monoclonal
Clone OTI2B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene C18orf54
Gene Alias LAS2
Gene Symbols C18orf54
Host Species Mouse
Immunogen Full length human recombit protein of human C18orf54 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162681
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.