Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C19orf10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559766
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559766 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559766 Supplier Invitrogen™ Supplier No. PA559766
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSEPTTVAFD VRPGGVVHSF SHNVGPGDKY TCMFTYASQG GTNEQWQMSL GTSEDHQHFT CTIWRPQGKS Y Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 87%.

The interleukins are a broad family of well characterized cytokines, primarily of hematopoietic cell origin. Cytokines are small, soluble proteins with pleiotropic effects on a variety of cell types. Cytokines have a regulatory function over the immune system and mediate aspects of inflammatory response. IL-25 (also called SF20/IL-25) is a secreted bone marrow stroma-derived growth factor cytokine that is derived from Th2 T-cells. IL-25 belongs to the IL-17 cytokine family and is capable of amplifying allergic type inflammatory responses by its actions on other cell types. IL-25 binds to mouse thymic shared antigen-1 and supports lymphoid cell proliferation. Infusion of mice with IL-25 induces IL-4, IL-5, and IL-13 gene expression. IL-25 induced mice also develop epithelial cell hyperplasia, increased mucus secretion, and airway hyperreactivity, suggesting that IL-25 may be an important mediator of allergic disease via production of IL-4, IL-5, IL-13 and eotaxin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C19orf10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYDGF
Gene Accession No. Q969H8
Gene Alias C19orf10; CS010; D17Wsu104e; EUROIMAGE1875335; IL 25; IL 27; Il25; IL27; IL27w; interleukin 25; interleukin 27 working designation; Interleukin25; interleukin-25; LOC501282; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; R33729 1; R33729_1; R337291; SF20; similar to lymphocyte antigen 6 complex, locus E ligand; stromal cell-derived growth factor SF20; UPF0556 protein C19orf10; UPF0556 protein C19orf10 homolog
Gene Symbols MYDGF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C19orf10. Recombinant protein control fragment (Product #RP-98886).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56005
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.