Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C19orf12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144268
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144268 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144268 Supplier Invitrogen™ Supplier No. PA5144268
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C19orf12 Polyclonal Antibody for ELISA, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen C19orf12
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene C19orf12
Gene Accession No. Q9NSK7
Gene Alias C19orf12; chromosome 19 open reading frame 12; NBIA3; NBIA4; neurodegeneration with brain iron accumulation 3; protein C19orf12; SPG43
Gene Symbols C19orf12
Host Species Rabbit
Immunogen Recombinant Human Protein C19orf12 protein (65-104AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83636
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.