Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C19orf174 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | C19orf174 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
C19orf174 Polyclonal specifically detects C19orf174 in Human samples. It is validated for Western Blot.Specifications
| C19orf174 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| BDAG1, BehcetS Disease Associated Gene 1, C9ORF174, CCDC180, Chromosome 9 open reading frame 174, Coiled-Coil Domain Containing 180, KIAA1529 | |
| The immunogen is a synthetic peptide directed towards the middle region of Human C19orf174 (NP_065944). Peptide sequence QKWMHSLPNDWIMENPVLHREKERAKREKARESENTIAAREVRGLMDTIV | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 100499483 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title