Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C19orf20 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735216 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5216 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5216 Supplier Proteintech Group Inc Supplier No. 261131AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TPGS1 is a protein coding gene.

Specifications

Antigen C19orf20
Applications Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TPGS1
Gene Accession No. Q6ZTW0, Q99MS8
Gene Alias C19orf20, GTRGEO22, PGs1
Gene Symbols TPGS1
Host Species Rabbit
Immunogen C19orf20 Fusion Protein Ag23546
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110012, 91978
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.