Learn More
Invitrogen™ C19orf21 Polyclonal Antibody

Description
Immunogen sequence: VLDGDTSYTY HLVCMGPEAS GWGQDEPQTW PTDHRAQQGV QRQGVSYSVH AYTGQPSPRG LHSENREDEG WQVYR Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 48%.
Specifications
Specifications
| Antigen | C19orf21 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MISP |
| Gene Accession No. | Q8IVT2 |
| Gene Alias | 9130017N09Rik; C19orf21; caprice; MISP; MISP1; mitotic interactor and substrate of Plk1; mitotic spindle positioning; mitotic spindle positioning protein |
| Gene Symbols | MISP |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.