Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C19orf22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570962
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570962 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570962 Supplier Invitrogen™ Supplier No. PA570962
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Human: 100%; Mouse: 93%; Pig: 100%; Rat: 100%; Zebrafish: 79%.

R3HDM4 gene ontology annotations related to this gene include nucleic acid binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C19orf22
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene R3HDM4
Gene Accession No. Q96D70
Gene Alias AI503502; C030046I01Rik; C19orf22; C20H19orf22; C7H19orf22; MGC137387 protein; R3H domain containing 4; R3H domain-containing protein 4; R3H domain-containing protein C19orf22; R3H domain-containing protein C19orf22 homolog; R3HDM4; RGD1307067; Unknown (protein for IMAGE:7291161); zgc:158644
Gene Symbols R3HDM4
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human C19orf22.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91300
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.