Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C19orf24 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561404
Description
Immunogen sequence: AGPCSLASWM LSQPGRGSQV KTGGTPTATA QDAEAPLPDC DLCLSPAPVG TWQPRAKAGW AGDPRNLSGN TFSP Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 28%.

Specifications
C19orf24 | |
Polyclonal | |
Unconjugated | |
C19orf24 | |
C19orf24; chromosome 19 open reading frame 24; FAM174C; Protein FAM174C; uncharacterized membrane protein C19orf24 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55009 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BVV8 | |
C19orf24 | |
Recombinant protein corresponding to Human C19orf24. Recombinant protein control fragment (Product #RP-99009). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction