Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C19orf24 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989255
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9255 Supplier Bioss Supplier No. BS9679R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen C19orf24
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C19orf24
Gene Accession No. Q9BVV8
Gene Alias C19orf24; chromosome 19 open reading frame 24; FAM174C; Protein FAM174C; uncharacterized membrane protein C19orf24
Gene Symbols C19orf24
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C19orf24.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55009
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.