Learn More
C19ORF33 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305739AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
IMUP is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cytosol; nucleus; plasma membrane.Specifications
C19ORF33 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9GZP8 | |
C19orf33 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
C19orf33 | |
H2RSP, IMUP, IMUP-1, IMUP-2, HAI-2 related small protein, HAI-2-related small protein, hepatocyte growth factor activator inhibitor type 2-related small protein, immortalization up-regulated protein, immortalization-upregulated protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64073 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.