Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C19ORF33 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571125
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1125 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-1125 Supplier Bethyl Laboratories, Inc Supplier No. A305740AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

IMUP is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cytosol; nucleus; plasma membrane.

Specifications

Antigen C19ORF33
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene C19orf33
Gene Accession No. Q9GZP8
Gene Alias H2RSP, IMUP, IMUP-1, IMUP-2, HAI-2 related small protein, HAI-2-related small protein, hepatocyte growth factor activator inhibitor type 2-related small protein, immortalization up-regulated protein, immortalization-upregulated protein
Gene Symbols C19orf33
Host Species Rabbit
Immunogen Between 56 and 106
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64073
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.