Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C19orf35 Monoclonal Antibody (3-B7-G3)
GREENER_CHOICE

Catalog No. PIMA532871
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532871 Supplier Invitrogen™ Supplier No. MA532871
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C19orf35 Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen C19orf35
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 3-B7-G3
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% Glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene PEAK3
Gene Accession No. Q6ZS72
Gene Alias C19orf35; chromosome 19 open reading frame 35; PEAK3; putative uncharacterized protein C19orf35; Uncharacterized protein PEAK3
Gene Symbols PEAK3
Host Species Mouse
Immunogen Recombinant protein within Human C19orf35 aa 1-473.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374872
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.