Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C19orf39 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562669
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562669 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562669 Supplier Thermo Scientific Supplier No. PA562669
Add to Cart
Add to Cart

C19orf39 Polyclonal Antibody

C19orf39 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen C19orf39
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SWSAP1
Gene Accession No. Q6NVH7
Gene Alias 2310047B19Rik; ATPase SWSAP1; C19orf39; SWIM type zinc finger 7 associated protein 1; SWIM-type zinc finger 7 associated protein 1; SWIM-type zinc finger 7-associated protein 1; SWS1AP1; SWS1-associated protein 1; Swsap1; zinc finger, SWIM-type containing 7 associated protein 1; ZSWIM7AP1; ZSWIM7-associated protein 1
Gene Symbols SWSAP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C19orf39. Recombinant protein control fragment (Product #RP-99179).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126074
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.