Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C19orf43 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563805
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563805 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563805 Supplier Invitrogen™ Supplier No. PA563805
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSTLSFVGKR RGGNKLALKT GIVAKKQKTE DEVLTSKGDA WAKYMAEVKK YKAHQCGDDD KTR Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.

C19orf43, also named as MGC2803 and fSAP18, is a protein coding gene. Exoribonuclease that is part of the telomerase RNA 3' end processing complex and which has the ability to all four unpaired RNA nucleotides from 5' end or 3' end with higher efficiency for purine bases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C19orf43
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Trir
Gene Accession No. Q9BQ61
Gene Alias 1110002H14Rik; 2310036O22Rik; AI854688; C19orf43; chromosome 19 open reading frame 43; Exoribonuclease Trir; fSAP18; My029 protein; RGD1564093; RIKEN cDNA 2310036O22 gene; similar to RIKEN cDNA 2310036O22; Telomerase RNA component interacting RNase; Trir; uncharacterized protein C19orf43; uncharacterized protein C19orf43 homolog
Gene Symbols Trir
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C19orf43. Recombinant protein control fragment (Product #RP-101247).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79002
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.