Learn More
Invitrogen™ C19orf43 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144072
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human MCF-7 whole cell, human 293T whole cell, human HepG2 whole cell. IHC: human liver cancer tissue, human tonsil tissue, human lung cancer tissue, human intestinal cancer tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
C19orf43, also named as MGC2803 and fSAP18, is a protein coding gene. Exoribonuclease that is part of the telomerase RNA 3' end processing complex and which has the ability to all four unpaired RNA nucleotides from 5' end or 3' end with higher efficiency for purine bases.
Specifications
| C19orf43 | |
| Polyclonal | |
| Unconjugated | |
| Trir | |
| 1110002H14Rik; 2310036O22Rik; AI854688; C19orf43; chromosome 19 open reading frame 43; Exoribonuclease Trir; fSAP18; My029 protein; RGD1564093; RIKEN cDNA 2310036O22 gene; similar to RIKEN cDNA 2310036O22; Telomerase RNA component interacting RNase; Trir; uncharacterized protein C19orf43; uncharacterized protein C19orf43 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79002 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q9BQ61 | |
| Trir | |
| A synthetic peptide corresponding to a sequence of human TRIR (AKYMAEVKKYKAHQCGDDDK). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.