Learn More
Invitrogen™ C19orf44 Polyclonal Antibody

Description
Immunogen sequence: SRNLTKIAPG HSRFLKRNQT LDEKHLLLKE NPVLGSGPRL ASCRPPTTAS RIRANAALMK LAQLETRIMN RKLQRNLSDT ESDSMT Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 54%.
Specifications
Specifications
| Antigen | C19orf44 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | C19orf44 |
| Gene Accession No. | Q9H6X5 |
| Gene Alias | C19orf44; chromosome 19 open reading frame 44; FLJ21742; RGD1311847; similar to 1700030K09Rik protein; uncharacterized protein C19orf44; Uncharacterized protein C19orf44 homolog |
| Gene Symbols | C19orf44 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.