Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C19orf45 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570168
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570168 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570168 Supplier Invitrogen™ Supplier No. PA570168
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Dog: 86%; Human: 100%; Pig: 86%.

TEX45 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C19orf45
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TEX45
Gene Accession No. Q8NA69
Gene Alias 1700019B03Rik; C19orf45; C20H19orf45; C2H19orf45; chromosome 19 open reading frame 45; RGD1563425; RIKEN cDNA 1700019B03 gene; similar to FLJ35784 protein; testis expressed 45; testis-expressed protein 45; TEX45; uncharacterized protein C19orf45; uncharacterized protein C19orf45 homolog
Gene Symbols TEX45
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human C19orf45.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374877
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.