Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C19orf45 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989257
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9257 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9257 Supplier Bioss Supplier No. BS9681R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TEX45 is a protein coding gene.

Specifications

Antigen C19orf45
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TEX45
Gene Accession No. Q8NA69
Gene Alias C19orf45; C20H19orf45; chromosome 19 open reading frame 45; Testis-expressed protein 45; TEX45; uncharacterized protein C19orf45; zinc finger protein 358; ZNF358
Gene Symbols TEX45
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C19orf45.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374877
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.