Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C19orf47 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | C19orf47 |
---|---|
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
C19orf47 Polyclonal antibody specifically detects C19orf47 in Human samples. It is validated for Western BlotSpecifications
C19orf47 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
chromosome 19 open reading frame 47, DKFZp686P05129, FLJ36888, hypothetical protein LOC126526 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human C19orf47. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
126526 | |
IgG | |
Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title