Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C19orf48 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559814
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559814 Supplier Invitrogen™ Supplier No. PA559814
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTVLEAVLEI QAITGSRLLS MVPGPARPPG SCWDPTQCTR TWLLSHTPRR RWISGLPR Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 40%.

The C19orf48 gene, located on chromosome 19q13, encodes a protein involved in various biological processes, including its role as a minor histocompatibility antigen recognized by CD8+ T cells. This gene has been implicated in oncogenesis due to its elevated expression in certain cancer types, such as breast cancer, where it has been associated with poor prognosis. The non-synonymous single-nucleotide polymorphisms within C19orf48 contribute to differential T-cell recognition in donor and recipient cells, highlighting its significance in immune responses. Additionally, C19orf48 has been linked with SNORD88C, a small nucleolar RNA (snoRNA) that guides methylation of rRNA, further illustrating its involvement in cellular proliferation and metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C19orf48
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C19orf48
Gene Accession No. Q6RUI8
Gene Alias C19orf48; chromosome 19 open reading frame 48; multidrug resistance-related protein; Uncharacterized protein C19orf48
Gene Symbols C19orf48
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C19orf48. Recombinant protein control fragment (Product #RP-99147).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84798
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.