Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C19orf52 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118310
This item is not returnable.
View return policy
Description
C19orf52 Polyclonal Antibody for Western Blot, ICC/IF, IP
TIMM29, also known as C19orf52, is identified as a novel subunit of the human TIM22 complex, which plays a critical role in the mitochondrial inner membrane. This protein is crucial for the stability of the TIM22 complex, which is responsible for the translocation of hydrophobic inner mitochondrial membrane proteins carrying internal targeting signals. TIMM29/C19orf52 is integrated into the mitochondrial inner membrane, exposing its C-terminus to the intermembrane space, and is vital for mitochondrial function and protein import pathways. This gene's importance is underscored by its involvement in processes related to mitochondrial integrity and its potential role in neurodegenerative disorders and viral interactions, such as those with the hepatitis B virus (HBV).
Specifications
| C19orf52 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9BSF4 | |
| TIMM29 | |
| E. coli-derived Human C19orf52/TIMM29 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| TIMM29 | |
| 1810026J23Rik; C19orf52; chromosome 19 open reading frame 52; Mitochondrial import inner membrane translocase subunit Tim29; RGD1309188; RIKEN cDNA 1810026J23 gene; similar to hypothetical protein BC011833; TIM29; TIMM29; uncharacterized protein C19orf52; uncharacterized protein C19orf52 homolog | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 90580 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction