Learn More
Invitrogen™ C19orf54 Polyclonal Antibody

Description
Immunogen sequence: HLVTGHPLLI PYDEDFNHEP CQRKGHKAHW AVSAGVLLGV RAVPSLGYTE DPELPGLFHP VLGTPCQPPS LP Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
Specifications
Specifications
| Antigen | C19orf54 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | C19orf54 |
| Gene Accession No. | Q5BKX5 |
| Gene Alias | C19orf54; chromosome 19 open reading frame 54; UPF0692 protein C19orf54 |
| Gene Symbols | C19orf54 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.