Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C19orf55 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569801
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 92%.
PROSER3 is a protein coding gene.
Specifications
C19orf55 | |
Polyclonal | |
Unconjugated | |
PROSER3 | |
C10H19orf55; C19orf55; C1H19orf55; LOW QUALITY PROTEIN: proline and serine-rich protein 3; proline and serine rich 3; proline and serine-rich protein 3; PROSER3; RGD1565712; uncharacterized protein C19orf55 homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
148137 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q2NL68 | |
PROSER3 | |
synthetic peptide directed towards the middle region of human C19orf55. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction