Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C19orf57 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560588
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560588 Supplier Invitrogen™ Supplier No. PA560588
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQIQDALDAS DFEAPPEQLF PSGNKPGPCW PGPSSHANGD PVAVAKAQPS RLIMGTHRDL EAFKRLNYRK TKLGGKAPLP YPSKGPGNIP RGDPPWREL Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 47%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C19orf57
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C19orf57
Gene Accession No. Q0VDD7
Gene Alias C19orf57; chromosome 19 open reading frame 57; pre-T/NK cell associated protein (3B3); pre-T/NK cell-associated protein 3B3; Uncharacterized protein C19orf57
Gene Symbols C19orf57
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C19orf57. Recombinant protein control fragment (Product #RP-99095).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79173
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.