Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C19orf61 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103357
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103357 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103357 Supplier Invitrogen™ Supplier No. PA5103357
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CS061.

This gene encodes a T cell adaptor protein, a class of intracellular molecules with modular domains capable of recruiting additional proteins but that exhibit no intrinsic enzymatic activity. The encoded protein contains a unique N-terminal region followed by a PH domain and C-terminal SH3 domain. Along with the adhesion and degranulation-promoting adaptor protein, the encoded protein plays a critical role in inside-out signaling by coupling T-cell antigen receptor stimulation to the activation of integrins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C19orf61
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SMG9
Gene Accession No. Q5PQS6, Q9DB90, Q9H0W8
Gene Alias 1500002O20Rik; AI429693; C19orf61; F17127_1; HBMS; N28092; protein SMG9; Protein SMG-9; protein smg-9 homolog; RGD1309888; Smg9; smg-9 homolog, nonsense mediated mRNA decay factor (C. elegans); SMG9 nonsense mediated mRNA decay factor; SMG9, nonsense mediated mRNA decay factor
Gene Symbols SMG9
Host Species Rabbit
Immunogen A synthesized peptide derived from human SMG9(Accession Q9H0W8), corresponding to amino acid residues V111-A161.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 365215, 56006, 71997
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.