Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C19orf61 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559866
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559866 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559866 Supplier Invitrogen™ Supplier No. PA559866
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KHSIKLVDDQ MNWCDSAIEY LLDQTDVLVV GVLGLQGTGK SMVMSLLSAN TPEEDQRTYV FRAQSAEMKE RGGNQTSGID FFITQERIVF L Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene encodes a T cell adaptor protein, a class of intracellular molecules with modular domains capable of recruiting additional proteins but that exhibit no intrinsic enzymatic activity. The encoded protein contains a unique N-terminal region followed by a PH domain and C-terminal SH3 domain. Along with the adhesion and degranulation-promoting adaptor protein, the encoded protein plays a critical role in inside-out signaling by coupling T-cell antigen receptor stimulation to the activation of integrins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C19orf61
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMG9
Gene Accession No. Q9H0W8
Gene Alias 1500002O20Rik; AI429693; C19orf61; F17127_1; HBMS; N28092; protein SMG9; Protein SMG-9; protein smg-9 homolog; RGD1309888; Smg9; smg-9 homolog, nonsense mediated mRNA decay factor (C. elegans); SMG9 nonsense mediated mRNA decay factor; SMG9, nonsense mediated mRNA decay factor
Gene Symbols SMG9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C19orf61. Recombinant protein control fragment (Product #RP-98994).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56006
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.