Learn More
Invitrogen™ C19orf63 Polyclonal Antibody

Description
C19orf63 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | C19orf63 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | EMC10 |
| Gene Accession No. | Q5UCC4 |
| Gene Alias | 2310044H10Rik; 5430410O10Rik; C19orf63; EMC10; ER membrane protein complex subunit 10; hematopoietic signal peptide-containing membrane domain-containing 1; hematopoietic signal peptide-containing membrane domain-containing protein 1; hematopoietic signal peptide-containing secreted 1; HSM1; HSS1; Inm02; Mirta22; UNQ764/PRO1556; UPF0510 protein INM02 |
| Gene Symbols | EMC10 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.