Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C19orf66 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559815
Description
Immunogen sequence: KFHGKASSKK AGALMRKFGS DHTGVGRSIV YGVKQKDGQE LSNDLDAQDP PEDMKQDRDI QAVATSLLPL TEANLRMFQR AQDD Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Inhibits programmed -1 ribosomal frameshifting (-1PRF) of a variety of mRNAs from viruses, such as HIV1, and cellular genes, such as PEG10. Interacts with the -1PRF signal of target mRNA and translating ribosomes and causes premature translation termination at the frameshifting site (PubMed:30682371). Regulates HIV1 GAG-POL expression by inhibiting -1PRF (PubMed:30682371). Exhibits antiviral activity against dengue virus (DENV) and can inhibit the replication of all DENV serotypes. May block the protein translation of DENV RNA via its association with cellular mRNA-binding proteins and viral RNA. Can also limit the replication of hepatitis C virus (HCV), West Nile virus (WNV), Chikungunya virus (CHIKV), herpes simplex virus type 1 (HHV-1) and human adenovirus (PubMed:26735137, PubMed:27974568). Binds nucleic acids with a higher affinity for ssRNA and ssDNA than for dsDNA (PubMed:27974568). [UniProt].
Specifications
C19orf66 | |
Polyclonal | |
Unconjugated | |
SHFL | |
C19orf66; chromosome 19 open reading frame 66; FLJ11286; Interferon-regulated antiviral protein; IRAV; Repressor of yield of Dengue virus; repressor of yield of DENV; repressor of yield of DENV protein; RYDEN; SFL; SHFL; Shiftless antiviral inhibitor of ribosomal frameshifting protein; UPF0515 protein C19orf66 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55337 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.25 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NUL5 | |
SHFL | |
Recombinant protein corresponding to Human C19orf66. Recombinant protein control fragment (Product #RP-99101). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction