Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C19orf67 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144807
Description
Immunogen sequence: DSLVDYYFLC YRDTWEDTGQ SPANSCPQIQ KLWSIGRWVP LGPAEDDLYS WILCPQPLGD YQQLLTIGFE EPTPTLATDL LVQILTGQ Antigen sequence identity: Mouse 89%; Rat 88%.
C19orf67 is a protein coding gene.
Specifications
C19orf67 | |
Polyclonal | |
Unconjugated | |
C19orf67 | |
C19orf67; chromosome 19 open reading frame 67; UPF0575 protein C19orf67; UPF0575 protein C19orf67-like | |
Rabbit | |
Affinity Chromatography | |
RUO | |
646457 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NJJ6 | |
C19orf67 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 251-338. Recombinant protein control fragment (Product #RP-110005). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction