Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C19ORF80 Monoclonal Antibody (OTI5A4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA808736
Description
The Betatrophin protein, also known as the hepatocellular carcinoma-associated protein TD26, is an adipocyte- and liver-enriched insulin targeted gene that is thought to play a role in lipid metabolism be regulating serum triglyceride levels. Betatrophin is closely related to the angiopoietin-like (ANGPTL) protein family. Betatrophin mRNA increases in the cold environment and after treatment with a high-fat diet, while its mRNA levels decrease in response to fasting, suggesting that it may play a role in mediating thermogenesis and energy homeostasis.Specifications
| C19ORF80 | |
| Monoclonal | |
| Unconjugated | |
| ANGPTL8 | |
| C19orf80, PRO1185, PVPA599, RIFL, TD26 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55908 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI5A4 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6UXH0 | |
| ANGPTL8 | |
| Human recombit protein fragment corresponding to amino acids 22-198 of human C19ORF80 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction