Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585571
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585571 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585571 Supplier Invitrogen™ Supplier No. PA585571
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (90%), Rat (91%), Chicken (80%), Rhesus Monkey (98%), Bovine (97%). Positive Control: Jurkat, Raji, NCI-H929. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a DNA binding and apoptosis-inducing protein and is localized in the nucleus. It is also a Rac3-interacting protein which acts as a corepressor for the thyroid hormone receptor. This protein is thought to regulate TRAX/Translin complex formation. Alternate splicing results in multiple transcript variants that encode the same protein. Multiple pseudogenes of this gene are found on chromosome 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1D
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene C1D
Gene Accession No. O35473, Q13901
Gene Alias 1110036E10Rik; AI875855; C1D; C1D DNA-binding protein; C1D nuclear receptor corepressor; C1D nuclear receptor co-repressor; hC1D; LRP1; mC1D; MGC12261; MGC14659; MGC188504; nuclear DNA binding protein; nuclear DNA binding protein, pseudogene 1; nuclear DNA-binding protein; nuclear nucleic acid-binding protein C1D; rCG23324; RGD1560600; Rrp47; small unique nuclear co-repressor; small unique nuclear receptor corepressor; small unique nuclear receptor co-repressor; SUNCOR; Sun-CoR
Gene Symbols C1D
Host Species Rabbit
Immunogen Full length human C1D Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10438, 57316
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.