Learn More
Description
Specifications
Specifications
| Antigen | C1GalT1 |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 5B6 |
| Conjugate | Unconjugated |
| Formulation | Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. |
| Host Species | Mouse |
| Immunogen | Recombinant fragment corresponding to the Lumenal region of human C1GalT1. |
| Purification Method | Protein G purified |
| Quantity | 25 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.