Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1GALT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552814
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552814 Supplier Invitrogen™ Supplier No. PA552814
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VDTQPNVLHN DPHARHSDDN GQNHLEGQMN FNADSSQHKD ENTDIAENLY QKVRILCWVM TGPQNLEKKA KHVKATWAQR CNKVLFMSSE ENKDFPAVGL KTKEGRDQLY WKTIK Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

Glycosyltransferase that generates the core 1 O-glycan Gal-beta1-3GalNAc-alpha1-Ser/Thr (T antigen), which is a precursor for many extended O-glycans in glycoproteins. Plays a central role in many processes, such as angiogenesis, thrombopoiesis and kidney homeostasis development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1GALT1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C1GALT1
Gene Accession No. Q9NS00
Gene Alias 2210410E06Rik; AV284120; B3Gal-T8; Beta-1,3-galactosyltransferase; C1GALT; C1GALT1; Core 1 beta1,3-galactosyltransferase 1; core 1 beta3-Gal-T; core 1 beta3-Gal-T1; core 1 O-glycan T-synthase; Core 1 synthase; core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1; core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1; core 1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 1; core1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase (C1galt1); core1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 1; core1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 13-galactosyltransferase (C1galt1); glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1; Plt1; T-syn; T-synthase
Gene Symbols C1GALT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C1GALT1. Recombinant protein control fragment (Product #RP-90065).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56913
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.